To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:IL-1β
- Clonality:Monoclonal
- Clone:B122
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:IL-1β
- Flow cytometry:Yes
- Size:50ug
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:IFNβ-inducing factor|Catabolin|IL-1b|IL1b il-1beta|Interleukin-1 beta
- Cat. no.:75843-594
- Supplier No.:82212-25-50
Specifications
About this item
The B122 monoclonal antibody specifically reacts with the precursor and mature secreted forms of mouse IL-1β and is cross-reactive with rat IL-1β. IL-1α and IL-1β are recognized by the same cell surface receptor but are generated from distinct genes. The B122 antibody is reported to neutralize the bioactivity of natural or recombinant IL-1β.
Type: Primary
Antigen: IL-1β
Clonality: Monoclonal
Clone: B122
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse