To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interferon gamma
- Clonality:Monoclonal
- Clone:NIB42
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:IFNG
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Type II interferon|T cell interferon|Interferon-g|Macrophage-activating factor (MAF)|IFNg|IFN-g|Interferon-γ|Immune interferon
- Cat. no.:75843-496
- Supplier No.:80821-25-500
Specifications
About this item
The NIB42 monoclonal antibody specifically reacts with human interferon gamma, a multifunctional cytokine that is produced mainly by activated T and NK cells. Interferon gamma has anti-viral and anti-parasitic properties and plays an important role in the activation, growth, and differentiation of macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-α, to inhibit proliferation of normal and transformed cells. The NIB42 antibody neutralizes IFN-γ.
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: NIB42
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human