To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:HLA-DR
- Clonality:Monoclonal
- Clone:LN3
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:HLA-DR
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:Major Histocompatibility Class II|MHC class II
- Cat. no.:75842-516
- Supplier No.:74111-80-100
Specifications
About this item
The LN3 monoclonal antibody specifically reacts with the human leukocyte antigen-DR (HLA-DR), a MHC class II heterodimeric surface glycoprotein. HLA-DR is expressed on the antigen presenting surfaces of monocytes, macrophages, dendritic cells, activated T cells, and B cells. It is integral to peptide presentation to CD4+ T cells.
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: LN3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human