To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:GARP (GARP5)
- Clonality:Monoclonal
- Clone:GARP5
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:GARP
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:LRRC32|Glycoprotein A repetitions predominant|Garpin
- Cat. no.:75842-530
- Supplier No.:77111-50-25
Specifications
About this item
The GARP5 monoclonal antibody specifically reacts with human Glycoprotein A Repetitions Predominant (GARP, also known as LRRC32 or Garpin) molecule, a 80 kDa transmembrane glycoprotein expressed on the cell surface. GARP is expressed in activated Treg cells, placenta, pancreas, liver, skeletal muscle, heart, lung, and kidney, and it acts as a receptor for latent TGF-β, which down-regulates the Treg cells action.
Type: Primary
Antigen: GARP
Clonality: Monoclonal
Clone: GARP5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human