To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Foxp3 (3G3)
- Clonality:Monoclonal
- Clone:3G3
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:Foxp3
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Zinc finger protein JM2|JM2|Forkhead box protein P3|Scurfin|IPEX
- Cat. no.:75842-606
- Supplier No.:83412-80-100
Specifications
About this item
The 3G3 monoclonal antibody specifically reacts with the mouse 50-55 kDa Foxp3 protein (JM2, IPEX), a member of the forkhead family of transcription factors. Foxp3 is expressed by the Treg lymphocytes, whose development and function are influenced by the forkhead protein. Ectopic expression of Foxp3 in T lymphocytes inhibits their activity and cytokine expression.Mutations of Foxp3 result in the “scurfy†mice phenotype.
Type: Primary
Antigen: Foxp3
Clonality: Monoclonal
Clone: 3G3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse