To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fc epsilon Receptor I alpha (FceR1) (MAR-1)
- Clonality:Monoclonal
- Clone:42795
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:FceR1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:FceRIa|high affinity IgE receptor
- Cat. no.:75842-668
- Supplier No.:84112-50-100
Specifications
About this item
The Mar-1 monoclonal antibody binds to the Fc ε Receptor I α subunit (FceR1a), which is a transmembrane glycoprotein from the immunoglobulin superfamily. FceR1a lacks signal-transducing ability and is expressed by mast and basophil cells.The Fc ε Receptor I α subunit is upregulated by IgE and forms a tetramer with a beta subunit and two gamma subunits, which have ITAM (immunoreceptor tyrosine-based activation motifs). The complex formed by the four subunits has very important roles in the IgE-facilitated allergic reactions.
Type: Primary
Antigen: FceR1
Clonality: Monoclonal
Clone: 42795
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse