To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:F4/80 antigen
- Clonality:Monoclonal
- Clone:BM8.1
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:F4/80
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:500 μg
- Form:liquid
- Antigen synonyms:Ly71|EMR1
- Cat. no.:75843-752
- Supplier No.:02922-50-500
Specifications
About this item
The BM8.1 monoclonal antibody specifically binds to the Mouse125 kDa F4/80 antigen, expressed by most mature macrophages. F4/80 is a transmembrane protein used as a marker of macrophages, although it is also expressed on Kupffer and Langerhans cells. The expression of F4/80 antigen is upregulated on bone marrow cells stimulated in vitro with the macrophage colony stimulating factor. The F4/80 antigen is a requirement for the induction of CD8 T cells-mediated peripheral tolerance.
Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: BM8.1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse