To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Thy-1
- Clonality:Monoclonal
- Clone:G7
- Host:Rat
- Isotype:IgG2c kappa
- Reactivity:Mouse
- Antigen symbol:CD90
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Thy-1 cell surface antigen|CDw90
- Cat. no.:75841-476
- Supplier No.:03032-25-500
Specifications
About this item
The G7 monoclonal antibody specifically reacts with mouse CD90, also known as thymus cell antigen-1 (Thy-1). CD90 is a 25-35 kDA GPI-anchored protein is the smallest member of the Ig superfamily. CD90 is expressed on thymocytes, neurons, hematopoietic stem cells, and T cells. It plays a role in T cell signal transduction and adhesion. The G7 antibody is reported to stimulate IL-2 secretion and T cell proliferation.
Type: Primary
Antigen: CD90
Clonality: Monoclonal
Clone: G7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2c, kappa
Reactivity: Mouse