To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD8 beta
- Clonality:Monoclonal
- Clone:H35-17.2
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CD8b
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Lyt-3|Ly-3|CD8 beta
- Cat. no.:75843-078
- Supplier No.:10132-60-100
Specifications
About this item
The H35-17.2 monoclonal antibody specifically reacts the mouse cd8 beta, a 32 kDa chain also known as Ly-3 and Lyt-3. CD8 is a T cell surface antigen co-receptor that interacts with MHC class I molecules on antigen presenting cells. It is expressed as a heterodimer with the alpha cd8 chain. The H35-17.2 antibody blocks T cell-mediated cytolysis of allogeneic lymphoma cells.
Type: Primary
Antigen: CD8b
Clonality: Monoclonal
Clone: H35-17.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse