To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of Differentiation 8a
- Clonality:Monoclonal
- Clone:OKT8
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:CD8a
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:T8|CD8 alpha|Ly-2|Lyt-2|Ly-35|Ly-B
- Cat. no.:75842-098
- Supplier No.:10121-77-25
Specifications
About this item
The OKT8 monoclonal antibody specifically reacts with human CD8α molecule, a tyPEI transmembrane glycoprotein of 32-34 kDa. CD8α is a member of the Ig superfamily, expressed as a homodimer (CD8αα) or as a heterodimer (CD8αβ). CD8+ αβ T lymphocytes express both CD8αα and CD8αβ, while some T lymphocytes and the natural killer cells express only the homodimers. CD8 binds to MHC class I and influences the development and the activation of T lymphocytes.OKT8, RPA-T8, and HIT8a antibodies do not compete with each other for binding to peripheral leukocytes, meaning that that they do not recognize the same epitoPEor block each other by steric hindrance.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: OKT8
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human