To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of Differentiation 8a
- Clonality:Monoclonal
- Clone:53-6.7
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:CD8a
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:T8|CD8 alpha|Ly-2|Lyt-2|Ly-35|Ly-B
- Cat. no.:75842-112
- Supplier No.:10122-25-500
Specifications
About this item
The 53-6.7 monoclonal antibody specifically reacts with Ly-2, the 38 kDa alpha chain, and with Lyt-2, the 34 kDa alpha’ chain, of the mouse CD8 antigen. The alpha’ chain is the truncated form of alpha chain, encoded by the same CD8a gene. In CD8a, the alpha and alpha’ chains form heterodimers with CD8b (the beta chains) or homodimers (alpha-alpha), which occur as receptors on the surface of the majority of thymocytes. A subpopulation of mature T lymphocytes expresses the CD8 alpha beta (alpha beta TCR T cells), and a subpopulation of intestinal intraepithelial lymphocytes and dendritic cells express CD8a without CD8b.CD8 interacts with the mouse major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells. Its function seems to be to attenuate the CD8-mediated signal for the stimulation of intrathymic T-cell maturation.The 53-6.7 antibody is useful for depleting CD8+ peripheral T lymphocytes. It cross reacts with the alpha- and alpha’-like polypeptides on some thymic and peripheral lymphocytes.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse