To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD83
- Clonality:Monoclonal
- Clone:Michel17
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Antigen symbol:CD83
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:HB15
- Cat. no.:75843-928
- Supplier No.:05912-60-100
Specifications
About this item
The Michel-17 monoclonal antibody specifically reacts with mouse CD83, a 45kDA type I transmembrane glycoprotein. CD83 is expressed mainly on activated lymphocytes and mature dendritic cells and it plays a role in T cell development. The Michel-17 antibody does not induce an activation signal on dendritic or T cells.
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: Michel17
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse