To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD81
- Clonality:Monoclonal
- Clone:EAT-2 (Eat2)
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:CD81
- Flow cytometry:Yes
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Tspan28|TAPA1|Tapa-1
- Cat. no.:75841-610
- Supplier No.:03912-25-100
Specifications
About this item
The Eat-2 monoclonal antibody reacts with mouse CD81, a 26kDA non-glycosylated transmembrane protein. CD81 is expressed broadly on B cells, T cells, NK cells, dendritic cells, fibroblasts, and endothelial cells. It plays a role in T cell development and is involved in B and T cell adhesion. The Eat-2 antibody is also reported to cross-react with the rat CD81 antigen.
Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: EAT-2 (Eat2)
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse