To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:AD2
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD73
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:Ecto-5'-nucleotidase|NT5E
- Cat. no.:75843-918
- Supplier No.:05811-80-25
Specifications
About this item
The AD2 monoclonal antibody specifically reacts with human CD73 (ecto-5’-nucleotidase), a 70 kDA glycosyl phosphatidylinositol (GPI)-anchored glycoprotein. CD73 catalyzes the dephosphorylation of adenosine monophosphate (AMP) to adenosine and is expressed on subsets of B, T, dendritic, endothelial, and mesenchymal stem cells. It is also involved in the adhesion of lymphocytes to endothelium and T cell activation.
Type: Primary
Antigen: CD73
Clonality: Monoclonal
Clone: AD2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human