To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD7 molecule
- Clonality:Monoclonal
- Clone:4H9
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:CD7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:gp40|Leu-9
- Cat. no.:75841-360
- Supplier No.:49111-50-100
Specifications
About this item
The 4H9 monoclonal antibody specifically binds to human CD7, a 40 kDa type I transmembrane glycoprotein known as Leu-9 or gp40. CD7 is found on the surfaces of T cells, NK cells, and acute lymphocytic leukemia cells. It is reported to be involved in modulating cytokine secretion and cellular adhesion.
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 4H9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human