To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD64
- Clonality:Monoclonal
- Clone:10.1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD64
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Fc gamma Receptor 1|FcR I|FcgR1|Fcgamma R1
- Cat. no.:75844-072
- Supplier No.:06711-25-500
Specifications
About this item
The 10.1 monoclonal antibody specifically reacts with human CD64, a 75kDA type I transmembrane glycoprotein that is the high affinity receptor for IgG. CD64 is expressed on macrophages and dendritic cells and is involved in antibody-dependent cellular toxicity, cytokine release phagocytosis, and endocytosis. The 10.1 antibody is reported to recognize the EC3 epitope of CD64.
Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: 10.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human