To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:P-selectin
- Clonality:Monoclonal
- Clone:Psel.KO2.3
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:SELP
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:LECAM-3
- Cat. no.:75841-644
- Supplier No.:04721-60-25
Specifications
About this item
The Psel.KO2.3 monoclonal antibody specifically reacts with mouse CD62P (P-Selectin), a 140 kDA type I transmembrane glycoprotein also known as granule membrane protein 140 (GMP-140). Upon activation it is quickly transported to the plasma membrane from the alpha-granules of platelets and WEibel-Palade bodies of endothelial cells. It is essential to the functions of cell adhesion during inflammatory reactions and in the interaction of platelets with monocytes and neutrophils. The Psel.KO2.3 antibody does not block adhesion and binds to an epitope common to several species including mouse, human, and rat.
Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: Psel.KO2.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human