To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:P-selectin
- Clonality:Monoclonal
- Clone:AK-4
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:SELP
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:LECAM-3
- Cat. no.:75843-816
- Supplier No.:04711-60-100
Specifications
About this item
The AK-4 monoclonal antibody specifically reacts with human CD62P (P-Selectin), a 140 kDA type I transmembrane glycoprotein also known as platelet activation-dependent granule membrane protein (PADGEM). Upon activation it is quickly transported to the plasma membrane from the alpha-granules of platelets and WEibel-Palade bodies of endothelial cells. It is essential to the functions of cell adhesion during inflammatory reactions and in the interaction of platelets with monocytes and neutrophils.
Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: AK-4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human