To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:L-selectin
- Clonality:Monoclonal
- Clone:MEL-14
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:CD62L
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:LAM-1|MEL-14|Ly-22|LECAM-1|L-selectin
- Cat. no.:75841-638
- Supplier No.:04712-80-100
Specifications
About this item
The MEL-14 monoclonal antibody specifically reacts with L- selectin (CD62L), a receptor with lectin-like and Epidermal Growth Factor-like domains. The weight of the CD62L molecules depend on their origin: 74 kDa (on lymphocytes) or 95 kDa (on neutrophils).In the Mouseorganism, CD62L can be expressed by most thymocytes, on subsets of B and T lymphocytes, monocytes, eosinophils, and neutrophils. The L-selectin binds sulfated, fucosylated, and glycosylated glycoproteins (MadCAM-1, GLYCAM-1, and CD 34). It mediates the migration of lymphocytes to the site of inflammation and their return to the peripheral lymphoid tissues and to the HEV (high endothelial venules). In vitro, L-selectin inhibits this binding and the lymphocyte extravasation into peripheral lymph nodes.
Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: MEL-14
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse