To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin beta 3
- Clonality:Monoclonal
- Clone:2C9.G3
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:ITGB3
- Flow cytometry:Yes
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:gpIIIa|integrin b3|integrin beta 3|CD61
- Cat. no.:75842-890
- Supplier No.:03712-25-100
Specifications
About this item
The 2C9.G3 monoclonal antibody specifically reacts with the mouse/rat CD61 molecule, known as the integrin beta 3 that forms the vitronectin receptor with CD51. The complex binds to other ligands such as fibrinogen, fibronectin, thrombospondin, and von WIllebrand factor. It is expressed by granulocytes, platelets, activated T cells, smooth muscle, and a subset of B cells.
Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: 2C9.G3
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse