To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin beta 3
- Clonality:Monoclonal
- Clone:VI-PL2
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:ITGB3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:gpIIIa|integrin b3|integrin beta 3|CD61
- Cat. no.:75842-886
- Supplier No.:03711-50-25
Specifications
About this item
The VI-PL2 monoclonal antibody reacts with CD61, a type I integral transmembrane glycoprotein also known as integrin beta 3. CD61 forms a complex with CD41 (platelet gpIIb) and CD51 (integrin alphaV). It is expressed on osteoclasts, fibroblasts, platelets, macrophages, and megakaryocytes and appears to bind to fibronectin, vitronectin, thrombospondin, and fibrinogen and vWF. The VI-PL2 antibody is reported to be cross-reactive with baboon, rhesus, and cynomolgus.
Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: VI-PL2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: mouse
Isotype: IgG1, kappa
Reactivity: Human