To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neural cell adhesion molecule
- Clonality:Monoclonal
- Clone:CMSSB
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:NCAM
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:CD56
- Cat. no.:75844-206
- Supplier No.:08611-60-100
Specifications
About this item
The CMSSB monoclonal antibody specifically reacts with human CD56, or Neural Cell Adhesion molecule (NCAM). CD56 is a 140 kDa transmembrane glycoprotein also known as Leu-19 or NKH1. It is expressed on NK and NKT cells and has a role in cellular adhesion.
Type: Primary
Antigen: NCAM
Clonality: Monoclonal
Clone: CMSSB
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human