To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD51/CD61
- Clonality:Monoclonal
- Clone:23C6
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD51/CD61
- Flow cytometry:Yes
- Size:50 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:ITGB3|ITGAV|vitronectin Receptor
- Cat. no.:75842-878
- Supplier No.:03611-25-50
Specifications
About this item
The 23C6 monoclonal antibody specifically reacts with the human CD51/61 molecule, known as the integrin alpha v Beta 3 complex or vitronectin receptor. The complex binds to other ligands such as CD29, fibrinogen, fibrinectin, vWf, laminin, and thrombospondin. It is expressed by endothelial cells, osteoclasts, melanoma cells, and at low levels on platelets and macrophages. The 23C6 antibody is reported to be able to block the receptors ability to form interactions with some of its ligands.
Type: Primary
Antigen: CD51/CD61
Clonality: Monoclonal
Clone: 23C6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human