To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:UCHT2
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD5
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:Ly-1|Leu-1|T1
- Cat. no.:75843-030
- Supplier No.:09111-60-100
Specifications
About this item
The UCHT2 monoclonal antibody specifically reacts with human CD5, a 67 kda type 1 transmembrane glycoprotein. CD5 is expressed on mature T cells, a subset of B cells, and peripheral blood dendritic cells. B cells that are CD5+ produce mostly IgM polyreactive antibodies. Its ligand is CD72, which is involved in T, B cell proliferation and interaction. The UCT2 antibody also recognizes non-human primate CD5.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human