To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:R1-2
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CD49d
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Integrin a 4 chain,|VLA4
- Cat. no.:75842-202
- Supplier No.:11522-25-500
Specifications
About this item
The R1-2 monoclonal antibody specifically reacts with mouse CD49d (integrin alpha 4), which forms the VLA-4 complex with CD29 (integrin beta 1). VLA-4 is expressed on thymocytes, monocytes, and a subset of peripheral lymphocytes. It is the receptor for fibronectin and CD106 (VCAM-1). CD49d also forms a receptor for mucosal vascular addressin (MAdCAM-1) with integrin beta 7.
Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: R1-2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse