To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HI100
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:CD45RA
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Cat. no.:75844-112
- Supplier No.:07121-50-25
Specifications
About this item
The HI100 monoclonal antibody specifically reacts with human CD45RA, the 220 kDa isoform of the human leukocyte common antigen (LCA) found on 40-50% of the peripheral CD4+ T lymphocytes, half of the peripheral CD8+ T lymphocytes and some of the monocytes and B lymphocytes. The CD45RA antigen is expressed by naïve and activated T lymphocytes. The HI100 monoclonal antibody is used as a phenotypic marker to discriminate T lymphocytes subsets.
Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: HI100
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human