To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase, receptor type, C
- Clonality:Monoclonal
- Clone:HI30
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:PTPRC
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Cat. no.:75844-096
- Supplier No.:07111-77-25
Specifications
About this item
The HI130 monoclonal antibody specifically reacts with the 180 kDa, 190 kDa, 205 kDa, and 220 kDa isoforms of the human leukocyte common antigen (LCA) CD45. It is expressed on lymphocytes, granulocytes, monocytes, thymocytes, and eosinophils, but not on mature erythrocytes, platelets, mature erythroid cells of bone marrow, and non-hematopoietic tissues. CD45 is essential for T cell activation and the tyrosine phosphatase activity of its intracellular region is integral for signal transduction.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI30
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human