To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase, receptor type, C
- Clonality:Monoclonal
- Clone:30-F11
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:PTPRC
- Conjugation:BG Violet 450
- Flow cytometry:Yes
- Size:25 ug
- Form:liquid
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Cat. no.:75841-966
- Supplier No.:07512-40-25
Specifications
About this item
The 30-F11 monoclonal antibody specifically reacts with all isoforms of CD45 and also with the alloantigens CD45.1 and CD45.2 (LCA). CD45 is a transmembrane glycoprotein, expressed by all the hematopoietic cells, except for platelets and mature erythrocytes, which distinguishes the leukocytes from the non-hematopoietic cells. The CD45 molecule is a member of the Protein Tyrosine Phosphatase (PTP) family, because its intracellular region contains two PTP domains. The extracellular region’s variability is caused by different levels of glycosylation, and the splicing of the 4, 5, and 6 exons.The isoforms found in the mouse strains depend on the activation state, maturation stage and cell type, and are very important in B and T lymphocytes antigen receptor signal transduction.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse