To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HIP1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD42b
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:gpIbalpha|gpIbα|Glycocalicin
- Cat. no.:75843-812
- Supplier No.:04511-50-100
Specifications
About this item
The HIP1 monoclonal antibody specifically binds to human CD42b, a 145kDa transmembrane protein known as the platelet glycoprotein Ib alpha chain (gpIbα). CD42 is expressed on platelets and megakaryocytes and is reported to be involved in platelet adhesion. The HIP1 antibody inhibits collage-induced aggregation and platelet to von Willebrand Factor (vWF) binding.
Type: Primary
Antigen: CD42b
Clonality: Monoclonal
Clone: HIP1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human