To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HIP8
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD41a
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:Integrin alpha Iib|platelet GPIIb
- Cat. no.:75842-860
- Supplier No.:03511-60-100
Specifications
About this item
The HIP8 monoclonal antibody specifically reacts with human CD41 (GPIIb), a subunit of the calcium-dependent complex with CD61 (IIIa). The complex is expressed on megakaryocytes and platelets. The HIP8 antibody blocks the aggregation of platelets and inhibits platelet activation.
Type: Primary
Antigen: CD41a
Clonality: Monoclonal
Clone: HIP8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human