To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:MWReg30
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:CD41
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:fibrinogen receptor
- Cat. no.:75842-872
- Supplier No.:03512-60-100
Specifications
About this item
The MWReg30 monoclonal antibody specifically reacts with mouse CD41, a transmembrane glycoprotein also known as integrin alpha 2b GPIIb. It is expressed on hematopoietic progenitors, megakaryocytes, and platelets. It forms a receptor with CD61 (integrin beta 3) that binds adhesion molecules such as fibrinogen, fibronectin, von Willebrand factor, and thrombin. Defects or absence of CD41 has been found to lead to coagulation disorders. The expression profile of CD150+,CD48-, and CD41- can be used to identify hematopoietic stem cells.
Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: MWReg30
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse