To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD40 Ligand
- Clonality:Monoclonal
- Clone:MR1
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:CD40LG
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:CD154
- Cat. no.:75842-298
- Supplier No.:16612-50-25
Specifications
About this item
The MR1 monoclonal antibody specifically reacts with mouse CD154, a 39kDA transmembrane glycoprotein also known as CD40 ligand and gp39. CD154 is expressed on activated T cells and plays a role in B-T cell costimulation. The interaction of CD154 with CD40 induces B cell cycle entry and the signaling for T-dependent B cell activation. By interfering with this interaction, the MR1 antibody is reported to inhibit T and B lymphocyte activation.
Type: Primary
Antigen: CD40LG
Clonality: Monoclonal
Clone: MR1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse