To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HM40-3
- Host:Armenian hamster
- Isotype:IgM kappa
- Reactivity:Mouse
- Antigen symbol:CD40
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:TNFRSF5
- Cat. no.:75843-686
- Supplier No.:02512-25-500
Specifications
About this item
The HM40-3 monoclonal antibody reacts with mouse CD40, a 48kDA type I glycoprotein known as Bp50. CD40 is part of the TNFR family and expressed on a subset of T cells, B cells, dendritic cells, and macrophages. It has been shown to be involved in the protection of B cells from apoptosis, B cell differentiation, co-stimulation, and isotype class-switching. The HM40-3 antibody blocks the binding of CD40L (CD154) and stimulates B and dendritic cell proliferation.
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: HM40-3
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgM, kappa
Reactivity: Mouse