To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:GK1.5
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CD4
- Conjugation:PE (Phycoerythrin)-Cy5®
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Leu-3|L3T4|T4
- Cat. no.:75843-974
- Supplier No.:06112-65-100
Specifications
About this item
The GK1.5 monoclonal antibody specifically binds with the mouse CD4 molecule, also known as L3T4, a 55 kDa differentiation antigen which binds to the MHC class II. CD4 is expressed on most thymocytes, a subpopulation of mature T lymphocytes, dendritic cells, pluripotent hematopoietic stem cells, B cell precursors, and lymphoid precursors inside the thymus. It is also expressed on the mouse egg cell membrane, enhancing adhesion to MHC class II bearing sperm. By interaction with MHC class II on the surface of APC, CD4 initiates the development of T lymphocytes and helps the optimum functioning of mature T lymphocytes. The binding of the GK1.5 antibody blocks the binding of the Anti-Mouse CD4 RM4-5 antibody.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse