To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:90
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:CD38
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:T10|ADP-ribosyl cyclase|Cyclic ADP-Ribose Hydrolase
- Cat. no.:75844-328
- Supplier No.:10412-25-500
Specifications
About this item
The 90 monoclonal antibody specifically bind to mouse CD38, a type II transmembrane glycoprotein. CD38 is expressed on activated T cells, plasma cells, monocytes, and macrophages. In the mouse as opposed to the human, CD38 expression is downregulated on mouse germinal center B cells and plasma cells. CD38 displays cyclase and hydrolase enzymatic activity and is involved in lymphocyte adhesion, activation, and cell signaling. Its ligand is CD31.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 90
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Mouse