To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:HIT2
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD38
- Conjugation:BG Violet 450
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:T10|ADP-ribosyl cyclase|Cyclic ADP-Ribose Hydrolase
- Cat. no.:75844-314
- Supplier No.:10411-40-100
Specifications
About this item
The HIT2 monoclonal antibody specifically bind to human CD38, a 45kda type II transmembrane glycoprotein. CD38 is expressed on activated T cells, plasma cells, monocytes, macrophages, dendritic cells, and thymocytes. CD38 displays cyclase and hydrolase enzymatic activity and is involved in lymphocyte adhesion, activation, and cell signaling. Its ligand is CD31.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human