To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tumor necrosis factor receptor superfamily member 18
- Clonality:Monoclonal
- Clone:DTA-1
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Antigen symbol:TNFRSF18
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:CD357|GITR|TNF receptor superfamily member 18|tumor necrosis factor receptor superfamily member 18|AITR|GITR-D
- Cat. no.:75843-366
- Supplier No.:39612-60-25
Specifications
About this item
The DTA-1 monoclonal antibody specifically reacts with mouse Glucocorticoid-Induced TNFR-related protein, also known as GITR and TNFRSF18, a 66-70 kDa homodimer glycoprotein, detected in the T cells treated with glucocorticoid dexamethasone. GITR is also expressed in naïve mice by CD25+/CD4+/CD8a- thymocytes and on CD25+/CD4+/CD45RB-low splenocytes. Low levels were detected in splenic CD25+/CD4+/CD45RB-low T cells, B cells, dendritic cells and macrophages. A GITR ligand was detected on dendritic cells, macrophages and B cells.The DTA-1 antibody stimulates GITR and abrogates suppression by T regulatory cells (Treg), without affecting their proliferation. DTA-1 administration or the removal of GITR-expressing cells led to organ specific autoimmune disease.
Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: DTA-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse