To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Jagged 1
- Clonality:Monoclonal
- Clone:HMJ1-29
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:JAG1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Ser-1|Htu|AWS|AHD|Jag1|JAGL1|jagged 1|ABE2|HJ1|Ozz|AGS|AGS1|Gsfabe2|CD339
- Cat. no.:75844-504
- Supplier No.:41412-60-100
Specifications
About this item
The HMJ1-29 monoclonal antibody reacts with mouse and rat CD339 (Jagged 1), a type 1 transmembrane Notch receptor ligand. It is expressed by macrophages and certain stromal, epithelial, dendritic, and thymic lymphoid cells. It has been reported to be play a role in Treg induction, Th2 polarization, neurogenesis, and cardiovascular development.
Type: Primary
Antigen: JAG1
Clonality: Monoclonal
Clone: HMJ1-29
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse