To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:WM53
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD33
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:Sialic Acid-Binding Immunoglobulin-Like Lectin 3|SIGLEC3
- Cat. no.:75843-912
- Supplier No.:05411-50-100
Specifications
About this item
The WM53 monoclonal antibody specifically reacts with human CD33, a 67 kDA transmembrane glycoprotein also known as GP67 and P67. CD33 is expressed on monocytes, granulocytes, dendritic cells, mast cells, and myeloid progenitors to the exclusion of hematopoietic stem cells, lymphocytes, and platelets. It functions to assist sialic acid-dependent cell adhesion.
Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: WM53
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human