To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:6C4
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD32
- Flow cytometry:Yes
- Size:50 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:FcGR2a|IGFR2
- Cat. no.:75843-804
- Supplier No.:04411-25-50
Specifications
About this item
The 6C4 monoclonal antibody specifically recognizes two isoforms of human CD32, Fc gamma RIIA and Fc gamma RIIB. From alternative mRNA splicing, 6 isoforms of CD32 have been reported. CD32 is a 40 kDA polymorphic transmembrane glycoprotein expressed on monocytes, macrophages, graulocytes, B cells, and platelets. The receptor has low affinity toward IgG and is involved in immunomodulation, phagocytosis, and the inflammation response. The 6C4 antibody in a rosette blocking assay is reported to inhibit Ig binding
Type: Primary
Antigen: CD32
Clonality: Monoclonal
Clone: 6C4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human