To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Platelet-Endothelial Cell Adhesion Molecule 1
- Clonality:Monoclonal
- Clone:390
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:PECAM1
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:endoCAM|PECAM-1|platelet and endothelial cell adhesion molecule 1|CD31|GPIIA|CD31/EndoCAM|PECA1
- Cat. no.:75842-850
- Supplier No.:03412-77-100
Specifications
About this item
The 390 monoclonal antibody specifically reacts with mouse cd31, a 130-140 kDA type I transmembrane glycoprotein also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1). CD31 is reported to bind to CD38 and is expressed on platelets, monocytes, granulocytes, and endothelial cells. It plays a role in angiogenesis, wound healing, cellular migration, and signal transduction.
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 390
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse