To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fetal liver kinase 1
- Clonality:Monoclonal
- Clone:Avas12a1
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:FLK1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Ly73|Ly-73|Flk-1|CD309|VEGF-R2
- Cat. no.:75843-374
- Supplier No.:41112-60-100
Specifications
About this item
The Avas12a1 monoclonal antibody specifically reacts with mouse CD309, also known as fetal liver kinase-1 (Flk-1) or the vascular endothelial growth factor receptor 2 (VEGFR2). CD309 is a receptor for VEGF and VEGFC and is expressed in endothelial cells in embryonic and adult tissue and a requirement for the development of vascular endothelial and hematopoietic cells.
Type: Primary
Antigen: FLK1
Clonality: Monoclonal
Clone: Avas12a1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse