To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:OKT3
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:CD3
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:Leu-4|CD3µ|T3
- Cat. no.:75842-910
- Supplier No.:05121-60-25
Specifications
About this item
The OKT3 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction.The OKT3 antibody is an immunosuppressive, which has proven to be an effective therapeutic agent in liver, heart, and renal allograft rejection.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human