To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:G4.18
- Host:Mouse
- Isotype:IgG3 kappa
- Reactivity:Rat
- Antigen symbol:CD3
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:T3
- Cat. no.:75841-690
- Supplier No.:05113-25-500
Specifications
About this item
The G4.18 monoclonal antibody specifically reacts with rat CD3, a T cell lineage marker and associated with the T cell receptor (TCR). CD3 is involved in antigen recognition, cell activation, and signal transduction and is found on subsets of NK-T, peripheral T, dendritic epidermal T, and thymocyte cells. Immobilized G4.18 antibody activates T cells in vitro and in vivo treatments of the G4.18 antibody prevents cardiac and skin allograft rejection.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: G4.18
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Rat