To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:UCHT1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD3
- Conjugation:Biotin
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:Leu-4|CD3µ|T3
- Cat. no.:75843-848
- Supplier No.:05131-30-25
Specifications
About this item
The UCHT1 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. Unlike HIT3a, another specific antibody of CD3, the UCHT1 antibody can stain both the surface and intracellular CD3ε. The immobilized UCHT1 can cross-link with the TCR complex, enhancing cellular activation and proliferation.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human