To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:17A2
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:CD3
- Conjugation:BG Violet 450
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:T3|T cell antigen receptor complex|CD3 epsilon
- Cat. no.:75842-894
- Supplier No.:05112-40-25
Specifications
About this item
The 17A2 monoclonal antibody specifically reacts with the mouse T lymphocytes receptor (TCR) associated CD3 complex, resulting in cellular activation and proliferation. CD3 is expressed by thymocytes and mature lymphocytes, and contains γ, δ, and ε subunits, involved in the assembly, trafficking, and surface expression of T-cell receptor complex. The interaction between the T lymphocytes and the 17A2 antibody can be blocked by the 145-2C11 anti-CD3e antibody, demonstrating that the 17A2 antibody recognizes the CD3 ε chain.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse