To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:SK7
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD3
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:500 tests
- Form:liquid
- Antigen synonyms:Leu-4|T3
- Cat. no.:75843-898
- Supplier No.:05141-80-500
Specifications
About this item
The SK7 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. The SK7 antibody is reported to have a mitogenic effect on the majority of peripheral blood T cells in the presence of functional monocytes.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: SK7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human