To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin beta 1
- Clonality:Monoclonal
- Clone:HMb1-1
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:ITGB1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:50 μg
- Form:liquid
- Antigen synonyms:Integrin Beta 1|Fibronectin Receptor Beta|CD29
- Cat. no.:75842-178
- Supplier No.:11312-60-50
Specifications
About this item
The HMb1-1 monoclonal antibody specifically reacts with mouse/rat CD29, a 130 kDA molecule also known as integrin beta 1, GPIIa, and the VLA-beta chain. It is expressed broadly on endothelial cells, epithelial, leukocytes, and smooth muscle. CD29 forms the VLA 1-6 complexes through the non-covalent interaction with the alpha integrins of CD49 a-f. The HMb1-1 antibody is capable of inhibiting T cell proliferation and blocking cell adhesion.
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: HMb1-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse