To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Toll-like receptor 2
- Clonality:Monoclonal
- Clone:TL2.1
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:TLR2
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 tests
- Form:liquid
- Antigen synonyms:TLR-2|CD282|toll-like receptor 2
- Cat. no.:75843-236
- Supplier No.:24911-60-100
Specifications
About this item
The TL2.1 monoclonal antibody specifically reacts with human CD282, the Toll-like receptor 2 (TLR2). It is a type I transmembrane signaling receptor containing IL-1 receptor like intracellular domain and leucine-rich repeats (LRR) in the extracellular domain. CD282 is expressed on peripheral blood monocytes and is involved distinguishing bacterial lipoproteins. The TL2.1 antibody blocks the receptor and immunoprecipitates TLR2 from human mammary epithelial cells (HMEC) and peripheral blood mononuclear cells (PBMC).
Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: TL2.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human