To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD278 (ICOS)
- Clonality:Monoclonal
- Clone:ISA-3
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:CD278
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:AILIM|H4|CRP-1
- Cat. no.:75842-392
- Supplier No.:30811-60-25
Specifications
About this item
The ISA-3 monoclonal antibody specifically reacts with human CD278 or Inducible COStimulatory molecule (ICOS), a 47-60 kDA homodimeric glycoprotein also known as H4, AILIM, and CRP-1. It is expressed on activated T cells and a group of thymocytes and is involved in the proliferation, activation, and cytokine production of T cells. When plate-bound, the ISA-3 antibody induces the production of IL-4,5,10 in T cells.
Type: Primary
Antigen: CD278
Clonality: Monoclonal
Clone: ISA-3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human